
cis-5,8,11,14,17-Eicosapentaenoic acid

Synonym(s):
EPA, Timnodonic acid
Linear Formula:
CH3(CH2CH=CH)5(CH2)3CO2H
CAS Number:
Molecular Weight:
302.45
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Tissue
Quality Segment
Functional Group
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing