Skip to Content
Merck
Ciprofloxacin European Pharmacopoeia (EP) Reference Standard

Ciprofloxacin

Ciprofloxacin European Pharmacopoeia (EP) Reference Standard
Synonym(s):
1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, Ciprobay
Empirical Formula (Hill Notation):
C17H18FN3O3
CAS Number:
Molecular Weight:
331.34

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique