Skip to Content
Merck
Chromium(III) chloride hexahydrate ≥96%

Chromium(III) chloride hexahydrate

Chromium(III) chloride hexahydrate ≥96%
Synonym(s):
Chromium trichloride hexahydrate, Hexaaquachromium(III) chloride
Linear Formula:
CrCl3 · 6H2O
CAS Number:
Molecular Weight:
266.45
EC Number:

Products

Product Category
Brand
Formula Weight
pH Value
Physical Form
Purity
Quality Segment
Reagent Type
Core
Special Grade