![Calcium levulinate dihydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/344/273/baebb6fd-6779-44a9-8c4e-c616a444c993/640/baebb6fd-6779-44a9-8c4e-c616a444c993.png)
Calcium levulinate dihydrate
![Calcium levulinate dihydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/344/273/baebb6fd-6779-44a9-8c4e-c616a444c993/640/baebb6fd-6779-44a9-8c4e-c616a444c993.png)
Synonym(s):
Calcium levulinate dihydrate, γ-Ketovaleric acid calcium salt, 4-Oxopentanoic acid calcium salt, Levulinic acid calcium salt dihydrate
Linear Formula:
CH3COCH2CH2COO · 1/2Ca · H2O
CAS Number:
Molecular Weight:
153.16
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing