
Calcium ionophore IV

Synonym(s):
N,N-Dicyclohexyl-N′,N′-dioctadecyl-3-oxapentanediamide, N,N-Dicyclohexyl-N′,N′-dioctadecyl-diglycolic diamide, ETH 5234
Empirical Formula (Hill Notation):
C52H100N2O3
CAS Number:
Molecular Weight:
801.36
Products
Product Category
Formula Weight
Melting Point (°C)
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing