![Beta-sitosterol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/422/879/af0013a3-bc33-441a-ad49-7d6c310ead83/640/af0013a3-bc33-441a-ad49-7d6c310ead83.png)
Beta-sitosterol
![Beta-sitosterol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/422/879/af0013a3-bc33-441a-ad49-7d6c310ead83/640/af0013a3-bc33-441a-ad49-7d6c310ead83.png)
Synonym(s):
β-Sitosterol, α-Dihydrofucosterol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol, 5-Stigmasten-3β-ol
Empirical Formula (Hill Notation):
C29H50O
CAS Number:
Molecular Weight:
414.71
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing