
β-Ecdysone

Synonym(s):
20-Hydroxyecdysone, β-Ecdysone, 2β,3β,14α,20β,22,25-Hexahydroxy-7-cholesten-6-one, Ecdysterone, Insect moulting hormone, Polypodine A
Empirical Formula (Hill Notation):
C27H44O7
CAS Number:
Molecular Weight:
480.63
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing