
β-D-Mannopyranose 1,3,4,6-tetra-O-acetate 2-O-trifluoromethanesulfonate

Synonym(s):
1,3,4,6-Tetra-O-acetyl-2-O-trifluoromethanesulfonyl-β-D-mannopyranose, Mannose triflate, TATM
Empirical Formula (Hill Notation):
C15H19F3O12S
CAS Number:
Molecular Weight:
480.36
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing