
Arecaidine hydrochloride

Synonym(s):
1,2,5,6-Tetrahydro-1-methylnicotinic acid
Empirical Formula (Hill Notation):
C7H11NO2 · HCl
CAS Number:
Molecular Weight:
177.63
Products
Formula Weight
Manufacturer
Product Line
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing