
Amiloxate

Synonym(s):
Isoamyl 4-methoxycinnamate, 4-Methoxycinnamic acid 3-methylbutyl ester, 4-Methoxycinnamic acid isoamyl ester, Amiloxate, Isopentyl 4-methoxycinnamate, NSC 408332
Empirical Formula (Hill Notation):
C15H20O3
CAS Number:
Molecular Weight:
248.32
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing