![Alumina NIST® SRM® 742, reference point](/deepweb/assets/sigmaaldrich/product/structures/242/244/fc2d9aa6-f301-4f00-96e0-a100dc2e9794/640/fc2d9aa6-f301-4f00-96e0-a100dc2e9794.png)
Alumina
![Alumina NIST® SRM® 742, reference point](/deepweb/assets/sigmaaldrich/product/structures/242/244/fc2d9aa6-f301-4f00-96e0-a100dc2e9794/640/fc2d9aa6-f301-4f00-96e0-a100dc2e9794.png)
Synonym(s):
Aluminum oxide, Alumina
Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing