
α-Hederin

Synonym(s):
(3β,4α)-3-[[2-O-(6-Deoxy-α-L-mannopyranosyl)-α-L-arabinopyranosyl]oxy]-23-hydroxyolean-12-en-28-oic acid
Empirical Formula (Hill Notation):
C41H66O12
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing