![Acridine Orange hydrochloride solution 10 mg/mL in H2O, ≥95.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/309/681/1e5bc32b-cbad-479e-b65c-f1aeed59ce11/640/1e5bc32b-cbad-479e-b65c-f1aeed59ce11.png)
Acridine Orange hydrochloride solution
![Acridine Orange hydrochloride solution 10 mg/mL in H2O, ≥95.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/309/681/1e5bc32b-cbad-479e-b65c-f1aeed59ce11/640/1e5bc32b-cbad-479e-b65c-f1aeed59ce11.png)
Synonym(s):
3,6-Bis(dimethylamino)acridine hydrochloride
Empirical Formula (Hill Notation):
C17H19N3 · HCl
CAS Number:
Molecular Weight:
301.81
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing