![Acebutolol hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/122/881/b3108702-cf82-4b0e-bef2-0ac62d354a2d/640/b3108702-cf82-4b0e-bef2-0ac62d354a2d.png)
Acebutolol hydrochloride
![Acebutolol hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/122/881/b3108702-cf82-4b0e-bef2-0ac62d354a2d/640/b3108702-cf82-4b0e-bef2-0ac62d354a2d.png)
Synonym(s):
Acebutolol hydrochloride, N-(3-Acetyl-4-[2-hydroxy-3-(isopropylamino)propoxy]phenyl)butanamide
Empirical Formula (Hill Notation):
C18H28N2O4 · HCl
CAS Number:
Molecular Weight:
372.89
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing