Skip to Content
Merck
4-tert-Octylphenol-monoethoxylate-ring-13C6 solution 10 μg/mL in acetone, analytical standard

4-tert-Octylphenol-monoethoxylate-ring-13C6 solution

4-tert-Octylphenol-monoethoxylate-ring-13C6 solution 10 μg/mL in acetone, analytical standard
Synonym(s):
2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethanol-13C6, 4-tert-OP1EO-13C6
Empirical Formula (Hill Notation):
13C6C10H26O2
CAS Number:
Molecular Weight:
256.33
EC Number:

Products

Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing