Skip to Content
Merck
4-tert-Octylphenol-diethoxylate-ring-13C6 solution 10 μg/mL in acetone, analytical standard

4-tert-Octylphenol-diethoxylate-ring-13C6 solution

4-tert-Octylphenol-diethoxylate-ring-13C6 solution 10 μg/mL in acetone, analytical standard
Synonym(s):
2-{2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethoxy}ethanol-13C6, 4-tert-OP2EO-13C6
Empirical Formula (Hill Notation):
13C6C12H30O3
CAS Number:
Molecular Weight:
300.38

Products

Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing