Skip to Content
Merck
4-Methylthiazole analytical standard

4-Methylthiazole

4-Methylthiazole analytical standard
Empirical Formula (Hill Notation):
C4H5NS
CAS Number:
Molecular Weight:
99.15
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing