
4-Hydroxyindole

Synonym(s):
4-Indolol, 4-Hydroxyindole
Empirical Formula (Hill Notation):
C8H7NO
CAS Number:
Molecular Weight:
133.15
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing