
4,4′-Dihydroxybiphenyl

Synonym(s):
4,4′-Biphenol, 4,4′-Biphenyldiol, 4,4′-Diphenol
Linear Formula:
HOC6H4C6H4OH
CAS Number:
Molecular Weight:
186.21
Products
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Monomer Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing