
4,4′-DDT solution

Synonym(s):
1,1,1-Trichloro-2,2-bis(4-chlorophenyl)ethane solution, 4,4′-DDT solution
Empirical Formula (Hill Notation):
C14H9Cl5
CAS Number:
Molecular Weight:
354.49
Products
Product Category
Brand
Formula Weight
Gene Alias
Product Line
Format
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing