Skip to Content
Merck
3-Methylbutyl 2-methylbutanoate ≥98%, FG

3-Methylbutyl 2-methylbutanoate

3-Methylbutyl 2-methylbutanoate ≥98%, FG
Synonym(s):
Isoamyl 2-methylbutyrate
Empirical Formula (Hill Notation):
C10H20O2
CAS Number:
Molecular Weight:
172.26
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing