
3′-Deoxy-3′-fluorothymidine

Empirical Formula (Hill Notation):
C10H13FN2O4
CAS Number:
Molecular Weight:
244.22
Products
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1