
2-Propanol-1,1,1,3,3,3-d6

Synonym(s):
Isopropanol-1,1,1,3,3,3-d6, Isopropyl-1,1,1,3,3,3-d6 alcohol
Linear Formula:
(CD3)2CHOH
CAS Number:
Molecular Weight:
66.13
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Isotope
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing