Skip to Content
Merck
2-Oxobutyric acid ≥95%, FG

2-Oxobutyric acid

2-Oxobutyric acid ≥95%, FG
Synonym(s):
2-Ketobutyric acid, α-Ketobutyric acid, Propionylformic acid, 2-Oxobutyric acid
Linear Formula:
CH3CH2COCOOH
CAS Number:
Molecular Weight:
102.09

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing