Skip to Content
Merck
2-Chloro-1,1,1-triethoxyethane 97%

2-Chloro-1,1,1-triethoxyethane

2-Chloro-1,1,1-triethoxyethane 97%
Synonym(s):
Triethyl 2-chloroorthoacetate, 2-Chloro-1,1,1-triethoxyethane
Empirical Formula (Hill Notation):
C8H17ClO3
CAS Number:
Molecular Weight:
196.67

Products

Product Category
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing