Skip to Content
Merck
2,7-Di-tert-butyl-9,9-dimethylxanthene-4,5-dicarboxylic acid 98%

2,7-Di-tert-butyl-9,9-dimethylxanthene-4,5-dicarboxylic acid

2,7-Di-tert-butyl-9,9-dimethylxanthene-4,5-dicarboxylic acid 98%
Empirical Formula (Hill Notation):
C25H30O5
CAS Number:
Molecular Weight:
410.50

Products

Product Category
Brand
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing