
24(S)-Hydroxycholesterol

Synonym(s):
24S-Hydroxycholesterol, 5-Cholesten-3β,24(S)-diol, Cerebrosterin, Cerebrosterol, Cholest-5-ene-3β,24-diol
Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing