
2,3-Dimethylaniline

Synonym(s):
1-Amino-2,3-dimethylbenzene, 2,3-Xylidine, 3-Amino-o-xylene, CN-Cbl, 2,3-Dimethylaniline
Linear Formula:
2,3-(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing