
1-Phenyl-1-trimethylsiloxyethylene

Synonym(s):
α-(Trimethylsiloxy)styrene, Acetophenone enol trimethylsilyl ether
Linear Formula:
C6H5C(=CH2)OSi(CH3)3
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing