Skip to Content
Merck
1-Octene analytical standard

1-Octene

1-Octene analytical standard
Synonym(s):
1-Octene
Linear Formula:
CH3(CH2)5CH=CH2
CAS Number:
Molecular Weight:
112.21

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Package Size
Reference Material Type
Special Grade
Technique