
1-Naphthyl phosphate monosodium salt monohydrate

Synonym(s):
α-Naphthyl phosphate monosodium salt monohydrate, Sodium 1-naphthyl phosphate monohydrate
Linear Formula:
C10H7OP(O)(OH)(ONa) · H2O
CAS Number:
Molecular Weight:
264.15
Products
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing