Merck
HomeProductsClinical & DiagnosticsProClin™ Preservatives
ProClin™ Preservatives
Used in over 1,000 FDA registered IVD kits from industry-leading diagnostics manufacturers, ProClin preservatives are water-soluble formulations that are among the most effective in the IVD industry. At low working concentrations, ProClin products help extend the shelf life of IVD reagents by effectively and immediately inhibiting a broad spectrum of microbes. ProClin preservatives are readily available in four unique formulations, ensuring a variety of options to meet a wide range of specific needs.
Products
Sort by Relevance
Showing 1-20 of 25
Change View
Showing 1-20 of 25
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-20 of 25 results
Page 1 of 2
Page 1 of 2
Proclin™ biocide preservative inhibits microbial Krebs cycle in several places
ProClin™ preservatives attack the Krebs cycle at four key points: the enzymes pyruvate dehydrogenase, α-ketoglutarate dehydrogenase, succinate dehydrogenase, and NADH dehydrogenase. Because all bacteria and fungi possess at least part of the Krebs cycle, they are broad spectrum in their activity.
ProClin™ preservatives are readily available in convenient pack sizes suitable for all manufacturing processes and volumes. Along side our four formulations, we also provide ProClin™ support products like a ProClin™ reference standard and a ProClin™ variety kit, useful in testing and evaluating the use of ProClin™ preservative within a specific diagnostic reagent or assay.
The ProClin™ Variety Kit (48119-U) contains 5 mL bottles of all four formulations and is an excellent tool for evaluating the suitability of ProClin™ products for their specific applications. See the characteristic differentiators between the formulations in our technical chart. Want to see how ProClin™ preservatives benefit your reagent? Request a sample.
Despite the enhanced safety profile of ProClin™ products, a study from an independent lab has shown equivalent or improved efficacy when compared to top alternatives like thimerosal and sodium azide. 
ProClin™ preservatives are restricted to use in IVD assay development and are intended for use in IVD manufacturing only. 
Disclaimer
The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.
We provide ProClin™ preservative product range with different characteristic formulations in multiple pack sizes to suit your specific needs.
Feature
ProClin™ 150
ProClin™ 200
ProClin™ 300
ProClin™ 950
Active Ingredient
(A.I.) %
CMIT/MIT
(1.5)
CMIT/MIT
(1.5)
CMIT/MIT
(3)
MIT
(9.5)
Stabilizer
23-25% Mg salts
3% Mg and Cu salts
Alkyl Carboxylate (salt-free)
None
Matrix
Water
Water
Modified glycol
Water
Working pH Range
2.5 - 8.5
2.5 - 8.5
2.5 - 8.5
2 - 12
Temperature Range
< 45 °C
< 45 °C
< 45 °C
< 90 °C
Typical Dosage Levels
(W/W)
0.06 - 0.10%
(9 – 15 ppm A.I.)
0.06 - 0.10%
(9 – 15 ppm A.I.)
0.03 - 0.05%
(9 – 15 ppm A.I.)
0.053 - 0.158%
(50 – 150 ppm A.I.)
Shelf Life
2 years
18 months
3 years
3 years
Related Resources
  • Brochure: ProClin™ Preservatives: Easy to incorporate into your diagnostic assay development
    ProClin™ preservatives are used in over 1,000 FDA registered IVD kits from industry leading manufacturers. At low working concentrations, ProClin™ preservatives can help extend the shelf life of IVD reagents by effectively and immediately inhibiting a broad spectrum of microbes
  • Flyer: For In Vitro Diagnostic Materials: Efficacy vs. Thimerosal and Sodium Azide
    The traditional preservatives used to prevent microbe growth in in vitro diagnostic products are not ideal. Thimerosal is expensive and, because it contains mercury, is classified as toxic for disposal.
  • FAQ: ProClin™ Preservatives Frequently Asked Questions
    ProClinTM products are dedicated for use in the Medical Devices (MD) and in vitro diagnostics (IVD) sector globally. The use of biocides to treat MD and IVD products is exempted from the Biocidal Products Regulation (BPR) as long as the MD/IVD products are in scope of the MD/IVD directives/regulations.
  • Technote: ProClin™ 300 Preservative
    ProClin™ 300 preservative is a highly effective biocide for the control of microorganisms in reagents and products intended for in vitro diagnostic use. Due to its broadspectrum activity, low toxicity at recommended use levels, excellent compatibility, and stability, ProClin™ 300 biocide is the ideal choice as an effective preservative in diagnostic reagents.