Skip to Content
Merck
All Photos(1)

Documents

G006149

UHPLC/MS Analysis of Various Drug Substances on Titan C18

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Titan C18, 5 cm x 2.1 mm I.D., 1.9 μm particles (577122-U)
mobile phase: [A] 5 mM ammonium formate; [B] 5 mM ammonium formate in 90:10 (v:v) acetonitrile:water
gradient: 30 to 70% B in 3.0 min
flow rate: 0.3 mL/min
column temp.: 35 °C
detector: MS/MS
injection: 3 μL
sample: 50 ng/mL in methanol

suitability

application for HPLC

application(s)

forensics and toxicology
pharmaceutical (small molecule)

Analysis Note

A Titan C18 column provided rapid, baseline resolution of this set of drug substances. The highest quality LC-MS solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.

Legal Information

Analyte

Description

    Metoprolol tartrate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    Propranolol hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Diazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service