Skip to Content
Merck
All Photos(1)

Key Documents

SML1788

Sigma-Aldrich

LJI308

≥98% (HPLC)

Synonym(s):

2,6-Difluoro-4-(4-(4-morpholinophenyl)pyridin-3-yl)phenol, 2,6-Difluoro-4-[4-[4-(4-morpholinyl)phenyl]-3-pyridinyl]phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18F2N2O2
CAS Number:
Molecular Weight:
368.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

OC1=C(C=C(C=C1F)C2=C(C=CN=C2)C3=CC=C(C=C3)N4CCOCC4)F

InChI

1S/C21H18F2N2O2/c22-19-11-15(12-20(23)21(19)26)18-13-24-6-5-17(18)14-1-3-16(4-2-14)25-7-9-27-10-8-25/h1-6,11-13,26H,7-10H2

InChI key

YUYJEQHNWKQNBT-UHFFFAOYSA-N

Biochem/physiol Actions

LJI308 is potent pan-RSK inhibitor that targets RSK N-terminal kinase domain (NTKD) ATP-binding site (IC50/[ATP] = 6 nM/5 μM/RSK1, 4 nM/20 μM/RSK2, 13 nM/10 μM/RSK3), exhibiting similary binding affinity toward RSK1-4, but much reduced inhibitory potency against S6K1 (IC50 = 0.8 μM), MEK4 (IC50 >10 μM), and HIP kinase 1 (IC50 >1 μM). LJI308 effectivelfy reduces cellular Y-box binding protein-1 (YB1) Ser102 (EC50 = 0.2-0.3 μM; MDA-MB-231 and H358 cells), but not ribosomal S6 protein (S6RP) S235/236, phosphorylation level and selectively inhibits the growth of YB1-overpressing HTRY-LT triple-negative breast cancer (TNBC) cells, but not the parental non-tumorigenic human mammary epithelial cell (HMEC) line HTRZ (% inhibition = 6.8%/HTRZ, 88%/HTRY-LT1, 66%/HTRY-LT2 in 8 days; 5 μM at 0 and 96 hr).
LJI308 is potent pan-RSK inhibitor that targets RSK N-terminal kinase domain ATP-binding site and inhibits YB1-dependent growth of cncer cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ida Aronchik et al.
Molecular cancer research : MCR, 12(5), 803-812 (2014-02-21)
The p90 ribosomal S6 kinase (RSK) family of serine/threonine kinases is expressed in a variety of cancers and its substrate phosphorylation has been implicated in direct regulation of cell survival, proliferation, and cell polarity. This study characterizes and presents the
Alastair H Davies et al.
Oncotarget, 6(24), 20570-20577 (2015-05-27)
The triple-negative breast cancer (TNBC) subtype is enriched in cancer stem cells (CSCs) and clinically correlated with the highest rate of recurrence. Several studies implicate the RSK pathway as being pivotal for the growth and proliferation of CSCs, which are
Rama Jain et al.
Journal of medicinal chemistry, 58(17), 6766-6783 (2015-08-14)
While the p90 ribosomal S6 kinase (RSK) family has been implicated in multiple tumor cell functions, the full understanding of this kinase family has been restricted by the lack of highly selective inhibitors. A bis-phenol pyrazole was identified from high-throughput
Mehdi Bouhaddou et al.
Cell, 182(3), 685-712 (2020-07-10)
The causative agent of the coronavirus disease 2019 (COVID-19) pandemic, severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), has infected millions and killed hundreds of thousands of people worldwide, highlighting an urgent need to develop antiviral therapies. Here we present a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service