Skip to Content
Merck
All Photos(1)

Documents

SML1505

Sigma-Aldrich

AZD1480

≥98% (HPLC)

Synonym(s):

(S)-5-Chloro-N2-(1-(5-fluoropyrimidin-2-yl)ethyl)-N4-(5-methyl-1H-pyrazol-3-yl)pyrimidine-2,4-diamine, 5-Chloro-N2-[(1S)-1-(5-fluoro-2-pyrimidinyl)ethyl]-N4-(5-methyl-1H-pyrazol-3-yl)-2,4-pyrimidinediamine, AZD-1480

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14ClFN8
CAS Number:
Molecular Weight:
348.77
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

InChI

1S/C14H14ClFN8/c1-7-3-11(24-23-7)21-13-10(15)6-19-14(22-13)20-8(2)12-17-4-9(16)5-18-12/h3-6,8H,1-2H3,(H3,19,20,21,22,23,24)/t8-/m0/s1

InChI key

PDOQBOJDRPLBQU-QMMMGPOBSA-N

Application

AZD1480 has been used as an inhibitor of janus kinase 2 (JAK2) and signal transducer and activator of transcription 3 (STAT3) in an in vitro cell model of upper gastrointestinal adenocarcinoma. It has also been used in combination with bortezomib to study tumor-associated macrophage-mediated survival of myeloma cells.

Biochem/physiol Actions

AZD1480 is an orally active, potent and selective inhibitor of Janus kinases JAK1 and JAK 2 with selectivity for JAK2 as evidenced by IC50 values of 1.3 nM for JAK1 and <0.4 nM for JAK2 in enzyme assays. AZD1480 inhibits endogenous as well as IL-6 induced STAT3 activation. AZD1480 reduces myeloid cell-mediated angiogenesis and metastasis. AZD1489 is an effective anticancer agent in bot adult and pediatric human tumors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AURKA regulates JAK2?STAT3 activity in human gastric and esophageal cancers.
Ahmed K, et al.
Molecular Oncology, 8(8), 1419-1428 (2014)
Tumour-associated macrophage-mediated survival of myeloma cells through STAT3 activation.
De Beule N, et al.
The Journal of Pathology, 241(4), 534-546 (2017)
Ilaria Marrocco et al.
Cells, 8(9) (2019-09-11)
Prostate cancer (PCa) is a multifactorial disease characterized by the aberrant activity of different regulatory pathways. STAT3 protein mediates some of these pathways and its activation is implicated in the modulation of several metabolic enzymes. A bioinformatic analysis indicated a
Namratha Sheshadri et al.
Cell death & disease, 12(11), 1038-1038 (2021-11-03)
Cancer cells experience endoplasmic reticulum (ER) stress due to activated oncogenes and conditions of nutrient deprivation and hypoxia. The ensuing unfolded protein response (UPR) is executed by ATF6, IRE1 and PERK pathways. Adaptation to mild ER stress promotes tumor cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service