Skip to Content
Merck
All Photos(1)

Documents

P5052

Sigma-Aldrich

5α-Pregnan-3α-ol-11,20-dione

powder, ≥98% (TLC)

Synonym(s):

3-Hydroxy-5α-pregnane-11,20-dione, Alfaxalone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

form

powder

drug control

USDEA Schedule IV

color

white

solubility

chloroform: ≥5 mg/mL, clear, colorless

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CC[C@H](C(C)=O)[C@@]4(C)CC(=O)[C@]3([H])[C@@]1(C)CC[C@@H](O)C2

InChI

1S/C21H32O3/c1-12(22)16-6-7-17-15-5-4-13-10-14(23)8-9-20(13,2)19(15)18(24)11-21(16,17)3/h13-17,19,23H,4-11H2,1-3H3/t13-,14+,15-,16+,17-,19+,20-,21+/m0/s1

InChI key

DUHUCHOQIDJXAT-OLVMNOGESA-N

Biochem/physiol Actions

Synthetic steroid compound alphaxalone (AFX) - 5α-Pregnan-3α-ol-11,20-dione- is a neurosteroid that directly activates and potentiates GABAA receptor-activated membrane current (IGABA). It acts as a positive allosteric modulator of the GABAA receptor; anesthetic. The alpha subunit subtype of GABAA determines the efficacy, but not the potency, of AFX neuroactive steroid to potentiate IGABA.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Maitra et al.
Brain research, 819(1-2), 75-82 (1999-03-20)
Neurosteroids are potent, endogenous modulators of GABAA receptor function in the central nervous system. The endogenous progesterone metabolite allopregnanolone (ALP) and the synthetic steroid compound alphaxalone (AFX) have been shown to both directly activate and potentiate GABAA receptor-activated membrane current
T Sooksawate et al.
Neuropharmacology, 37(9), 1103-1110 (1998-12-02)
The effects of increased membrane cholesterol on the potentiation of GABA(A) currents by pregnanolone (5beta-pregnan-3alpha-ol-20-one), allopregnanolone (5alpha-pregnan-3alpha-ol-20-one), alphaxalone (5alpha-pregnan-3alpha-ol-11,20-dione) and propofol were investigated in acutely dissociated rat hippocampal neurones using the whole-cell patch clamp technique. Cholesterol enrichment of the neurones

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service