Key Documents
OH30038553
Ohaus® ORP Electrode, STORP1
STORP1
About This Item
UNSPSC Code:
39121436
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30038553
ORP measuring range
-1000-+1000 mV
General description
All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
Application
pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement
Features and Benefits
Can be used in conjunction with all Starter bench and portable meters
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)