Skip to Content
Merck
All Photos(5)

Key Documents

HPA002042

Sigma-Aldrich

Anti-C1QTNF6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Complement C1q tumor necrosis factor-related protein 6 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

CQRCCDSEDPLDPAHVSSASSSGRPHALPEIRPYINITILKGDKGDPGPMGLPGYMGREGPQGEPGPQGSKGDKGEMGSPGAPCQKRFFAFSVGRKTALHSGEDFQTLLFERVFVNLDGCFDMATGQFAAPLR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

C1q and tumor necrosis factor related protein 6 (C1QTNF6) also known as CTRP6, is a member of C1QTNF family. It consists of a putative N-terminal signal peptide, short variable region, collagen like domain and C1q globular domain.

Immunogen

Complement C1q tumor necrosis factor-related protein 6 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-C1QTNF6 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

C1QTNF6 (C1q and tumor necrosis factor related protein 6) is mainly involved in the fatty acid metabolism. It stimulates the phosphorylation of acetyl coenzyme A carboxylase (ACC) and fatty acid oxidation in myocytes via the AMPK-ACC pathway. In addition, C1QTNF6 also facilitates the phosphorylation and activation of the 5′-AMP-activated protein kinase (AMPK) in skeletal muscle cells. It has also been reported that interleukin-10 expression is stimulated by C1QTNF6 in macrophages thus proving the existence of an anti-inflammatory property in the protein.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86364

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of a secretory protein C1qTNF6, a C1qTNF family member, in hepatocellular carcinoma.
Takeuchi, T., et al.
Analytical Cellular Pathology, 34, 113-121 (2011)
Wan Lee et al.
FEBS letters, 584(5), 968-972 (2010-01-28)
C1qTNF-related proteins (CTRPs) are involved in diverse processes including metabolism, inflammation host defense, apoptosis, cell differentiation, autoimmunity, hibernation, and organogenesis. However, the physiological role of CTRP6 remains poorly understood. Here we demonstrate that the globular domain of CTRP6 mediates the
Tamotsu Takeuchi et al.
Analytical cellular pathology (Amsterdam), 34(3), 113-121 (2011-04-22)
Recent studies have revealed that the adiponectin-associated protein belonging to the C1qTNF family mediates various biological processes. However, the pathobiological property of C1qTNF6 in carcinogenesis remains unclear. Here, we investigated the expression status of C1qTNF6 in human hepatocellular carcinomas and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service