Skip to Content
Merck
All Photos(3)

Key Documents

AB356480

Sigma-Aldrich

Anti-Cas9 (Rabbit Polyclonal)

serum, from rabbit

Synonym(s):

SaCas9, CRISPR-associated endonuclease Cas9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

Staphylococcus aureus, bacteria

packaging

antibody small pack of 25 μL

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

General description

CRISPR-associated endonuclease Cas9 (UniProt: J7RUA5; also known as SaCas9, Cas9) is encoded by the Cas9 gene in Streptococcus aureus. Cas9 belongs to subtype II-A subfamily with one HNH Cas9-type domain. CRISPR (clustered regularly interspaced short palindromic repeat) is an adaptive immune system that provides protection against mobile genetic elements (viruses, transposable elements and conjugative plasmids). CRISPR clusters contain spacers, sequences complementary to antecedent mobile elements, and target invading nucleic acids. Cas9 has a bi-lobed architecture with a recognition lobe (residues 41-425) and a discontinuous nuclease lobe (NUC, residues 1-40 and 453-1053); the crRNA-target DNA lies in a channel between the 2 lobes. The NUC lobe has 2 endonuclease domains. The discontinuous RuvC-like domain in NUC cleaves the target DNA non-complementary to crRNA while the HNH nuclease domain cleaves the target DNA complementary to crRNA. Cas9 serves as a RNA-guided DNA endonuclease that cleaves double-stranded DNA targets with a protospacer adjacent motif (PAM) and complementarity to the guide RNA. The homologous region of the Streptococcus pyogenes is not closely related in amino acid sequence in S. aureus, hence this antibody does not recognize that protein.

Specificity

This rabbit polyclonal antibody detects Cas9 from Staphylococcus aureus. It targets an epitope within 251 amino acids from the C-terminal end. It does not react with Cas9 from Streptococcus pyogenes.

Application

Anti-Cas9 (Rabbit Polyclonal), Cat. No. AB356480, is a highly specific rabbit polyclonal antibody that targets CRISPR-associated endonuclease Cas9 and has been tested in Immunofluorescence and Western Blotting.
Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected Cas9 in HEK293 cells with transfected with GFP-Cas9-SA (Courtesy of Dr. Gerry Shaw).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Cas9 in HEK293 cells overexpressing fusion protein GFP-Cas9 (Courtesy of Dr. Gerry Shaw).

Quality

Evaluated by Western Blotting in HEK293 cells overexpressing fusion protein GFP-Cas9.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Cas9 (Rabbit Polyclonal) in 40 µg of HEK293 cells overexpressing fusion protein GFP-Cas9 from Staphylococcus aureus.

Target description

~55 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zuriah Meacham et al.
PLoS biology, 21(12), e3002431-e3002431 (2023-12-08)
Bacteriophages encode anti-CRISPR (Acr) proteins that inactivate CRISPR-Cas bacterial immune systems, allowing successful invasion, replication, and prophage integration. Acr proteins inhibit CRISPR-Cas systems using a wide variety of mechanisms. AcrIIA1 is encoded by numerous phages and plasmids, binds specifically to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service