Skip to Content
Merck
All Photos(1)

Documents

566400

Sigma-Aldrich

Normal Guinea Pig Serum, Lyophilized Solid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.47

form

lyophilized solid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

wet ice

storage temp.

2-8°C

General description

Normal carrier sera, produced from pooled lots of sera from healthy, non-immunized guinea pig. Suitable for use in radioimmunoassay.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.25 M NaCl, 0.01 M Sodium Phosphate, pH 7.6.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Reconstitute in 5 ml of H₂O.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service