Skip to Content
Merck
All Photos(2)

Documents

492041

Sigma-Aldrich

Nrf2 Activator II, AI-1

The Nrf2 Activator II, AI-1, also referenced under CAS 75483-04-4, modulates the biological activity of Nrf2. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

Nrf2 Activator II, AI-1, Ethyl-4-chloro-1-methyl-2-oxo-1,2-dihydroquinoline-3-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12ClNO3
CAS Number:
Molecular Weight:
265.69
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C13H12ClNO3/c1-3-18-13(17)10-11(14)8-6-4-5-7-9(8)15(2)12(10)16/h4-7H,3H2,1-2H3

InChI key

XUJQDJGGHFNYDY-UHFFFAOYSA-N

General description

A cell-permeable chloro-quinolinone compound that selectively and covalently modifies Kelch-like ECH-associated protein 1 (Keap1)-Cys151 and represses Keap1 function and reduces Nrf2 (Nuclear factor E2-related factor 2)-Keap1 interaction. Shown to stabilize Nrf2 by inhibiting the Keap1-driven ubiquitination of Nrf2 and thereby activating the Nrf2-ARE (antioxidant response element) pathway (EC50 = 2.7 µM in neuroblastoma IMR-32 cells). Offers protection against H2O2 induced oxidative stress, but unlike tert-butylhydroquinone (Cat. No. 286888) AI-1 does not act as a free radical scavenger. Its anti-oxidant effects are mediated via Nrf2 induced antioxidant response. Does not affect mutant ARE where GC are substituted with AT within the core ARE sequence.
A cell-permeable chloro-quinolinone compound that selectively and covalently modifies Kelch-like ECH-associated protein 1 (Keap1)-Cys151 and represses Keap1 function and reduces Nrf2 (Nuclear factor erythroid 2-related factor 2)-Keap1 interaction. Shown to stabilize Nrf2 by inhibiting the Keap1-driven ubiquitination of Nrf2 and thereby activating the Nrf2-ARE (antioxidant response element) pathway (EC50 = 2.7 µM in neuroblastoma IMR-32 cells). Offers protection against H2O2 induced oxidative stress, but unlike tert-butylhydroquinone (Cat. No. 286888). AI-1 does not act as a free radical scavenger. Its anti-oxidant effects are mediated via Nrf2 induced antioxidant response. Does not affect mutant ARE where GC are substituted with AT within the core ARE sequence.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Keap1
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Hur, W., et al. 2010. Chem. Biol.17, 537.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wooyoung Hur et al.
Chemistry & biology, 17(5), 537-547 (2010-06-11)
Eukaryotic cells counteract oxidative and other environmental stress through the activation of Nrf2, the transcription factor that controls the expression of a host of protective enzymes by binding to the antioxidant response element (ARE). The electrophilic molecules that are able

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service