208731
Calpain Substrate II, Fluorogenic
Cyclic fluorogenic substrate for the quantitative determination of calpain -1 and -2.
Synonym(s):
Calpain Substrate II, Fluorogenic, Suc-LY-AMC
About This Item
Quality Level
Assay
≥95% (TLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated
protect from light
color
off-white
solubility
DMSO: soluble
fluorescence
λex ~380 nm
λem ~460 nm
shipped in
ambient
storage temp.
−20°C
InChI
1S/C29H33N3O8/c1-4-16(2)27(32-24(34)11-12-25(35)36)29(39)31-22(14-18-5-8-20(33)9-6-18)28(38)30-19-7-10-21-17(3)13-26(37)40-23(21)15-19/h5-10,13,15-16,22,27,33H,4,11-12,14H2,1-3H3,(H,30,38)(H,31,39)(H,32,34)(H,35,36)/t16?,22?,27-/m0/s1
InChI key
VSWNAJAOJZXWBM-FTJGSZSVSA-N
General description
Biochem/physiol Actions
Calpain -1 and -2
Warning
Sequence
Reconstitution
Other Notes
Woo, K.M., et al. 1989. J. Biol. Chem. 264, 2088.
Sasaki, T., et al. 1984. J. Biol. Chem.259, 12489.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service