Skip to Content
Merck
All Photos(1)

Key Documents

197217

Sigma-Aldrich

Bcl-xL BH4423, Human, Cell-Permeable

A cell-permeable peptide that prevents apoptotic cell death by directly binding to the voltage-dependent anion channel (VDAC) and blocking its activity.

Synonym(s):

Bcl-xL BH4423, Human, Cell-Permeable, HIV-TAT₄₈₋₅₇-β-Ala-Bcl-x L BH4₄₋₂₃, Ac-GRKKRRQRRR-βA-SNRELVVDFLSYKLSQKGYS-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C165H279N59O46
Molecular Weight:
3825.35
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

20% acetic acid: 5 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable peptide that prevents apoptotic cell death by directly binding to the voltage-dependent anion channel (VDAC) and blocking its activity. Leads to the inhibition of cytochrome c release and loss of mitochondrial membrane potential (ΔΨm). Contains the conserved N-terminal homology domain (BH4) of Bcl-xL (amino acids 4 - 23) that has been shown to be essential for inhibiting VDAC activity in liposomes and in isolated mitochondria. The BH4 domain is linked to a carrier peptide, a 10-amino acid HIV-TAT48-57 sequence with a β-alanine residue as a spacer for maximum flexibility. Following its uptake, it is mainly localized to the mitochondria.
A highly cell-permeable peptide that prevents apoptotic cell death by directly binding to the voltage-dependent anion channel (VDAC) and blocking its activity. Ultimately results in the inhibition of cytochrome c release and loss of mitochondrial membrane potential (ΔΨm). Contains the conserved N-terminal homology domain (BH4) of Bcl-xL (amino acids 4-23) that has been shown to be essential for inhibiting VDAC activity in liposomes and isolated mitochondria. The BH4 domain is linked to the carrier peptide, the 10-amino acid HIV-TAT48-57 sequence with a β-alanine residue as a spacer for maximum flexibility. Following its uptake, it is mainly localized in the mitochondria.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Voltage-dependent anion channel (VDAC)
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-βAla-Ser-Asn-Arg-Glu-Leu-Val-Val-Asp-Phe-Leu-Ser-Tyr-Lys-Ser-Gln-Lys-Gly-Tyr-Ser-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chen m., et al. 2002. J. Biol. Chem. 277, 29181.
Futaki, S., et al. 2001. J. Biol. Chem.276, 5836.
Shimizu, S., et al. 2000. Proc. Natl. Acad. Sci. USA97, 3100.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kensuke Kataoka et al.
Frontiers in cell and developmental biology, 8, 307-307 (2020-07-17)
Tendons and ligaments are pivotal connective tissues that tightly connect muscle and bone. In this study, we developed a novel approach to generate tendon/ligament-like tissues with a hierarchical structure, by introducing the tendon/ligament-specific transcription factor Mohawk (MKX) into the mesenchymal
Yi-Hsien Fang et al.
International journal of molecular sciences, 21(7) (2020-04-03)
Mature mammalian hearts possess very limited regenerative potential. The irreversible cardiomyocyte loss after heart injury can lead to heart failure and death. Pluripotent stem cells (PSCs) can differentiate into cardiomyocytes for cardiac repair, but there are obstacles to their clinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service