Skip to Content
Merck
All Photos(2)

Key Documents

1.14979

Supelco

Chlorine Test Refill pack

for use with MCOLORTEST®

Synonym(s):

Chlorine Visual Test Kit Refills

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

feature

refill

Quality Level

specific analyte(s)

chlorine

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

When the reagents of a MQuant® test (formally MColortest®) are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of test kits, making it a particularly economical and ecological system.
This refill pack contains all reagents for the MQuant® test kit 1.14978 for the colometric evaluation of chlorine.
for 1.14978 MQuant®

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

AQA Standard for free chlorine - Preparation of a standard solution for free chlorine

Preparation of a standard solution for free Chlorine according to DIN EN ISO 7393

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service