Skip to Content
Merck
All Photos(1)

Key Documents

N-067

Supelco

Naphyrone hydrochloride

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Naphyrone hydrochloride solution, Energy-1, NRG-1, O-2482

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23NO · HCl
CAS Number:
Molecular Weight:
317.85
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CCCC(N1CCCC1)C(C2=CC(C=CC=C3)=C3C=C2)=O.[H]Cl

InChI

1S/C19H23NO.ClH/c1-2-7-18(20-12-5-6-13-20)19(21)17-11-10-15-8-3-4-9-16(15)14-17;/h3-4,8-11,14,18H,2,5-7,12-13H2,1H3;1H

InChI key

SIOVYKZRZLWQSY-UHFFFAOYSA-N

General description

Also known as NRG-1, Energy-1, or O-2482, naphyrone is a new designer drug and stimulant with cases of abuse reported in Europe. The drug emerged in late 2010 as a new legal high in the UK after the banning of mephedrone. This certified solution standard offers numerous applications in GC/MS or LC/MS testing from forensic analysis and clinical toxicology to urine drug monitoring.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service