Skip to Content
Merck
All Photos(3)

Key Documents

A13409

Sigma-Aldrich

α-Acetylbutyrolactone

≥99%

Synonym(s):

2-Acetyl-γ-butyrolactone, 2-Acetylbutanolide, 2-Acetylbutyrolactone, 2-Oxo-3-acetyltetrahydrofuran, 3-Acetyl-4,5-dihydrofuran-2-one, 3-Acetyloxolan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O3
CAS Number:
Molecular Weight:
128.13
Beilstein:
112676
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

liquid

refractive index

n20/D 1.459 (lit.)

bp

107-108 °C/5 mmHg (lit.)

density

1.19 g/mL at 25 °C (lit.)

SMILES string

CC(=O)C1CCOC1=O

InChI

1S/C6H8O3/c1-4(7)5-2-3-9-6(5)8/h5H,2-3H2,1H3

InChI key

OMQHDIHZSDEIFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service