Merck
HPMA copolymer-phospholipase C and dextrin-phospholipase A2 as model triggers for polymer enzyme liposome therapy (PELT).
Journal of drug targeting (2017-07-22)
Elaine L Ferguson, Anna Scomparin, Hanna Hailu, Ronit Satchi-Fainaro
RÉSUMÉ
'Polymer Enzyme Liposome Therapy' (PELT) is a two-step anticancer approach in which a liposomal drug and polymer-phospholipase conjugate are administered sequentially to target the tumour interstitium by the enhanced permeability and retention effect, and trigger rapid, local, drug release. To date, however, the concept has only been described theoretically. We synthesised two polymer conjugates of phospholipase C (PLC) and A2 (PLA2) and evaluated their ability to trigger anthracycline release from the clinically used liposomes, Caelyx
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
3,3-Dimethylglutaric acid, 98%