Merck
Role of Megakaryocytes in Breast Cancer Metastasis to Bone.
Cancer research (2017-02-17)
Walter Jackson, Donna M Sosnoski, Sara E Ohanessian, Paige Chandler, Adam Mobley, Kacey D Meisel, Andrea M Mastro
RÉSUMÉ
Little is known about how megakaryocytes may affect metastasis beyond serving as a source of platelets. In this study, we explored the functional implications of megakaryocyte accumulation in the femurs of mice after injection of metastatic or non-metastatic breast cancer cells in 4T1.2 BALB/cJ and MDA-MB-231 nude mouse models. At bone metastatic sites, but not primary growth sites, tumor growth was associated with increased megakaryopoiesis in both model systems. In the orthotopic BALB/cJ model, extramedullary hematopoiesis occurred in the spleen, resulting in a four-fold increase in megakaryocytes. In support of the hypothesis that reducing megakaryocytes may reduce metastasis, we found that thrombopoietin-deficient mice exhibited a 90% relative decrease in megakaryocytes, yet they developed more aggressive metastasis than wild-type hosts. In human clinical specimens, we observed an increase in megakaryocytes in the bone marrow of 6/8 patients with metastatic breast cancer compared with age- and gender-matched controls. Taken together, our results suggested that an increase in megakaryocytes occurring in response to metastatic cells entering the bone marrow confers some measure of protection against metastasis, challenging present views on the role of megakaryocytes in this setting.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Human TPO / Thyroid Peroxidase ELISA Kit, for serum, plasma, cell culture supernatants and urine