Merck
Synthesis and activity of 3-pyridylamine ligands at central nicotinic receptors.
European journal of medicinal chemistry (2001-01-04)
G Balboni, M Marastoni, S Merighi, P Andrea Borea, R Tomatis
RÉSUMÉ
A series of thirty 2-(3-pyridylaminomethyl)azetidine, pyrrolidine and piperidine analogues as nicotinic acetylcholine receptor (nAChR) ligands was explored. In general, pyrrolidinyl and many azetidinyl compounds were found to bind with enhanced affinity relative to the piperidines. In the three series, the parallel structural changes (stereochemistry, N-methylation and/or chloro substitution) do not consistently lead to parallel shifts in affinity. The more active compounds (K(i) affinity values ranging from 8.9 to 90 nM) were about as analgesic as nicotine in a tail-flick assay in mice after subcutaneous injections.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
N-(tert-Butoxycarbonyl)-D-prolinal, 97%