Merck
  • Biotin-containing phospholipid vesicle layer formed on self-assembled monolayer of a saccharide-terminated alkyl disulfide for surface plasmon resonance biosensing.
Biotin-containing phospholipid vesicle layer formed on self-assembled monolayer of a saccharide-terminated alkyl disulfide for surface plasmon resonance biosensing.
Journal of bioscience and bioengineering (2008-06-19)
Yoshiko Ishizuka-Katsura, Tetsuichi Wazawa, Tadato Ban, Kenichi Morigaki, Shigeru Aoyama
RÉSUMÉ
We describe a technique to form a biotin-containing phospholipid vesicle layer on a self-assembled monolayer (SAM) deposited on a gold surface to immobilize biotinylated receptor proteins for a surface plasmon resonance (SPR) biosensor. The adsorption of vesicle of 1,2-dioleoyl-sn-glycero-3-phosphocholine (DOPC) was examined by SPR on the SAMs of dithiobis(1-deoxy-glucitol-1-carbamoyl pentane) (DDGP), 11-mercaptoundecanoic acid, 11-mercaptoundecanol, 11-amino-1-undecanethiol, and 12-mercaptododecane, and it was found that the DOPC vesicle rapidly adsorbed on the DDGP SAM to achieve the highest coverage of the surface. By quartz crystal microbalance with dissipation monitoring (QCM-D), the DOPC layer formed on the DDGP SAM was shown to be a vesicle layer, in which intact DOPC vesicles physisorbed on the SAM surface. To immobilize a biotinylated receptor protein, one of three biotinylated phospholipids, 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(biotinyl) (biotin-DOPE), N-((6-(biotinoyl)amino)hexanoyl)-1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine (biotin-X-DHPE) and N-(biotinoyl)-1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine (biotin-DHPE), was mixed with DOPC to form a biotin-containing vesicle layer on the DDGP SAM. A comparative binding study of NeutrAvidin and the biotin-containing vesicle layers showed that the use of biotin-X-DHPE achieved the most rapid immobilization of NeutrAvidin on the vesicle layer at the highest surface density. Furthermore, biotinylated protein A, as a receptor protein, could be immobilized through NeutrAvidin on the vesicle layer containing DOPC and biotin-X-DHPE, and its reaction with immunoglobulin G, as an analyte, was successfully observed by SPR. The results demonstrate that the biotin-containing vesicle layer on the DDGP SAM must be a useful component for SPR biosensor surfaces.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Avanti
18:1 Biotinyl PE, Avanti Research - A Croda Brand 870282P, powder
Sigma-Aldrich
SAM Formation reagent for biotin terminated monolayer on gold surface